missing translation for 'onlineSavingsMsg'
Learn More

ANKRD27 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1236.20 SEK - 3313.85 SEK

Specifications

Antigen ANKRD27
Concentration 0.29 mg/mL
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation
Conjugate Unconjugated
Format Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16812635
View Documents Promotion Details
Proteintech
24034-1-AP-150UL
150 μL
3313.85 SEK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 31-01-2022
Please to purchase this item. Need a web account? Register with us today!
16822635
View Documents Promotion Details
Proteintech
24034-1-AP-20UL
20 μL
1236.20 SEK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 31-01-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The product of this gene is a membrane-associated protein that colocalizes with huntingtin. This protein has similarities to cytoskeleton proteins and its interaction with huntingtin is thought to play a functional role in the cell filament network. Loss of normal huntingtin-HIP1 interaction in Huntington disease may contribute to a defect in membrane-cytoskeletal integrity in the brain. This gene could help in the understanding of the normal function of huntingtin and also the pathogenesis of Huntington disease. It also has been implicated in the pathogenesis of hematopoietic maligcies. An alternative splice variant of this gene has been described but its full length sequence has not been determined.
Specifications

Specifications

ANKRD27
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Polyclonal
-20°C
0.29 mg/mL
Unconjugated
Rabbit
Human
IgG
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications