missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD33 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ANKRD33 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823765
|
Proteintech
25202-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813765
|
Proteintech
25202-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ankyrin repeat domain 33 (ANKRD33), also named as C12orf7, belongs to the ankyrin repeat family and is highly expressed in retina. ANKRD33 exists as two isoforms. The isoform 1 molecular weight is 29 kDa and isoform 2 is 49 kDa. The function of ANKRD33 is still non-known.Specifications
ANKRD33 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q7Z3H0 | |
341405 | |
ANKRD33 Fusion Protein Ag18582 | |
Primary | |
-20°C | |
ANKRD33 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ANKRD33, ankyrin repeat domain 33, C12orf7 | |
ANKRD33 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |