missing translation for 'onlineSavingsMsg'
Learn More

ANKS3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA559589

Additional Details : Weight : 0.02000kg

Product Code. 15767441

  • 3670.00 SEK / 100µL


Estimated Shipment date
from Supplier 03-03-2020
Add to basket

Specifications

Specifications

ANKS3
Polyclonal
Unconjugated
ANKS3
Liquid
ANKS3
Recombit protein corresponding to Human ANKS3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q6ZW76
ankyrin repeat and SAM domain-containing protein 3
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
124401
Videos
Documents

Research Use Only