missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
Ankyrin 3 Polyclonal antibody specifically detects Ankyrin 3 in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifikationer
Specifikationer
| Antigen | Ankyrin 3 |
| Användningsområden | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Klassificering | Polyclonal |
| Konjugera | APC |
| Utspädning | Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence |
| Formulering | PBS |
| Gene Alias | ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier |
| Värd art | Chicken |
| Immunogen | The C-terminal 398 amino acids of human Ankyrin 3 expressed in and purified from E. coli. |
| Reningsmetod | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?