missing translation for 'onlineSavingsMsg'
Learn More

Annexin A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55018-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16836455

  • 1236.20 SEK / 20µL
Estimated Shipment: 19-05-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ANXA1 is a pleiotrophic protein produced by many cell types including peripheral blood leucocytes. The antibody is specific to ANXA1.

Annexin I belongs to a family of Ca (2+) -dependent phospholipid binding proteins that are preferentially located on the cytosolic face of the plasma membrane. Annexin I has a molecular mass of 40 kDa, with phospholipase A2 inhibitory activity. Since phospholipase A2 is required for the biosynthesis of the potent mediators of inflammation, prostaglandins and leukotrienes, annexin I may have potential anti-inflammatory activity.
Specifications

Specifications

Annexin A1
Polyclonal
Unconjugated
ANXA1
Annexin 1, annexin A1, Annexin I, ANX1, ANXA1, Calpactin 2, Calpactin II, Chromobindin 9, Lipocortin I, LPC1, p35
Rabbit
Antigen Affinity Chromatography
RUO
16952, 25380, 301
Antibody
IgG
Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P04083, P07150, P10107
Anxa1
Peptide
20 μL
Primary
Rat, Human, Mouse
Liquid
Videos
SDS
Documents
Certificates
Special Offers

Special Offers