missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A3 Mouse anti-Human, Clone: 1A11E4, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Annexin A3 |
---|---|
Clone | 1A11E4 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838255
|
Proteintech
66405-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16828255
|
Proteintech
66405-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.Specifications
Annexin A3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
35 alpha calcimedin, Annexin 3, annexin A3, Annexin III, ANX3, ANXA3, Lipocortin III, PAP III | |
ANXA3 | |
IgG2a | |
Protein A | |
Antibody |
1A11E4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P12429 | |
306 | |
Annexin A3 Fusion Protein Ag2381 | |
Primary | |
-20°C | |
ANXA3 |