missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin IV Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10087-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.Specifications
Annexin IV | |
Polyclonal | |
Unconjugated | |
ANXA4 | |
35 beta calcimedin, Annexin 4, annexin A4, Annexin IV, ANX4, ANXA4, Chromobindin 4, Endonexin I, Lipocortin IV, P32.5, PAP II, PIG28, PP4 X, Protein II, ZAP36 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11746, 307, 79124 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09525, P55260, P97429 | |
Anxa4 | |
Annexin IV Fusion Protein Ag0130 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |