Läs mer
STEMCELL Technologies Annexin V, FITC
Produktkod. 17147561
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 tester
25 tester
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17147561 25 tester
17157561 100 tester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17147561 Leverantör STEMCELL Technologies Leverantörsnummer 1000332
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Assay buffer for labeling with Annexin V
  • Annexin V is a member of the annexin family of proteins that bind to membrane phospholipids in the presence of calcium
  • This dye has high affinity for phosphatidylserine (PS) that is present in the inner leaflet of the plasma membrane
  • During early-stage cell apoptosis, PS is translocated from the inner to the outer leaflet of the plasma membrane, exposing it to the external environment
  • Annexin V, a characteristic marker for early cell apoptosis, detects the translocation of PS to the external environment
  • Annexin V is used along with viability dyes such as 7-AAD (Catalog #75001) or Propidium Iodide (Catalog #75002)
  • The process of PS translocation occurs prior to the loss of membrane integrity
  • Therefore, as cells progress through apoptosis and towards necrosis, the cell membrane is compromised and consequently, viability dyes pass into the cell
  • Thus, cells undergoing early apoptosis stain positive for Annexin V and negative for viability dyes, while apoptotic death or necrosis is characterized by positive staining for both Annexin V and the viability dye
Specifikationer
Kvantitet 25 tester
För användning med (applikation) Flow Cytometry
Konjugera FITC