missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
Calcium-activated chloride channels (CaCC) are present in many cell types and mediate physiological functions such as epithelial secretion, sensory signal transduction, and smooth muscle contraction. Subunits of these CaCCs include the transmembrane proteins TMEM16A and TMEM16B. TMEM16B is predicted to have eight transmembrane domains with both the amino and carboxy termini in the cytoplasm and is expressed in several tissues including olfactory sensory neurons as well as photoreceptors in mammalian retina. Like TMEM16A, TMEM16B is thought to form at least part of CaCCs but has different biophysical characteristics such as voltage dependence and unitary conductance.
Specifikationer
Specifikationer
| Antigen | ANO2 |
| Användningsområden | Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassificering | Polyclonal |
| Koncentration | 0.55 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gen | ANO2 |
| Genaccessionsnr. | Q8CFW1, Q9NQ90 |
| Gene Alias | ANO2, anoctamin 2, C12orf3, TMEM16B, Transmembrane protein 16B |
| Gensymboler | ANO2 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?