missing translation for 'onlineSavingsMsg'
Learn More

ANO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20647-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16880215

  • 3313.85 SEK / 150µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Calcium-activated chloride channels (CaCC) are present in many cell types and mediate physiological functions such as epithelial secretion, sensory signal transduction, and smooth muscle contraction. Subunits of these CaCCs include the transmembrane proteins TMEM16A and TMEM16B. TMEM16B is predicted to have eight transmembrane domains with both the amino and carboxy termini in the cytoplasm and is expressed in several tissues including olfactory sensory neurons as well as photoreceptors in mammalian retina. Like TMEM16A, TMEM16B is thought to form at least part of CaCCs but has different biophysical characteristics such as voltage dependence and unitary conductance.
Specifications

Specifications

ANO2
0.55 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CFW1, Q9NQ90
ANO2, anoctamin 2, C12orf3, TMEM16B, Transmembrane protein 16B
Rabbit
IgG
150 μL
-20°C
Primary
100361584, 243634, 57101
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ANO2
Liquid
ANO2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers