Learn More
ANP32A Recombinant Protein, Abnova
Brand:  Abnova H00008125-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16132292
  • 5030.41 SEK / 25µg
Estimated Shipment: 23-11-2021
Description
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH07200
Solution
8125
ANP32A (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
C15orf1/I1PP2A/LANP/MAPM/MGC119787/MGC150373/PHAP1/PHAPI/PP32
ANP32A
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
53.13
8
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ANP32A
Human
Yes
Videos
SDS
missing translation for 'documents'
Product Certifications