missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anthrone, MP Biomedicals™
Click to view available options
Quantity:
10 g
25 g
100 g
Specifications
Formula Weight | 194.22 |
Color | Yellow |
Quantity | 10 g |
Physical Form | Powder |
Chemical Name or Material | Anthrone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'