Läs mer
v-abl Abelson murine leukemia viral oncogene homolog 2 (arg, Abelson-related gene), Mouse, Clone: 5C6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ABL2.
Brand: Abnova H00000027-M09.100ug
Beskrivning
This gene encodes a member of the Abelson family of nonreceptor tyrosine protein kinase. The protein is highly similar to the ABL1 protein, including the tyrosine kinase, SH2 and SH3 domains, and has a role in cytoskeletal rearrangements by its C-terminal F-actin- and microtubule-binding sequences. This gene is expressed in both normal and tumor cells, and is involved in translocation with the ETV6 gene in leukemia. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene. [provided by RefSeq
Sequence: KKTLGLRAGKPTASDDTSKPFPRSNSTSSMSSGLPEQDRMAMTLPRNCQRSKLQLERTVSTSSQPEENVDRANDMLPKKSEESAAPSRERPKAKLLPRGASpecifikationer
| v-abl Abelson murine leukemia viral oncogene homolog 2 (arg, Abelson-related gene) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC065912 | |
| ABL2 | |
| ABL2 (AAH65912, 743 a.a. ∼ 842 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 5C6 | |
| Mouse monoclonal antibody raised against a partial recombinant ABL2. | |
| ABL2 | |
| ABLL/ARG/FLJ22224/FLJ31718/FLJ41441 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 27 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.