missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADCK1 Monoclonal Antibody (1E8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA522336
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: MARKALKLAS WTSMALAASG IYFYSNKYLD PNDFGAVRVG RAVATTAVIS YDYLTSLKSV PYGSEEYLQL RSKVHLRSAR RLCELCCANR GTFIKVGQHL
ADCK1 Monoclonal antibody specifically detects ADCK1 in Human samples. It is validated for ELISA.Specifications
ADCK1 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q86TW2 | |
ADCK1 | |
ADCK1 (AAH58906.1, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
1E8 | |
Unconjugated | |
ADCK1 | |
2610005A10Rik; aarF domain containing kinase 1; AarF domain-containing protein kinase 1; Adck1; Nrbf1; uncharacterized aarF domain-containing protein kinase 1 | |
Mouse | |
Affinity chromatography | |
RUO | |
57143 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |