missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenovirus Type 2 Monoclonal Antibody (4D2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511222
Additional Details : Weight : 0.01000kg
Description
MA5-11222 targets Adenovirus Fiber in IF and WB applications and shows reactivity with Virus samples. The MA5-11222 immunogen is uV-irradiated Ad2 virus.
Adenovirus Type 2 Monoclonal antibody specifically detects Adenovirus Type 2 in Virus samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Adenovirus Type 2 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a κ |
Immunocytochemistry, Western Blot | |
4D2 | |
Unconjugated | |
Adeno Virus; Adeno Virus 2; AdV; AdV-2; HADV; HADV-2 | |
UV-irradiated Ad2 virus | |
500 μL | |
Primary | |
4° C | |
Liquid |