missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1C4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518511
Additional Details : Weight : 0.01000kg
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols by utilizing NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme catalyzes the bioreduction of chlordecone, a toxic organochlorine pesticide, to chlordecone alcohol in liver. This gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14.Specifications
AKR1C4 | |
Polyclonal | |
Unconjugated | |
AKR1C4 | |
3-alpha-HSD; 3-alpha-HSD1; 3-alpha-hydroxysteroid dehydrogenase type I; AKR1C4; Aldo-keto reductase family 1 member C4; aldo-keto reductase family 1, member C4; C11; CDR; CHDR; Chlordecone reductase; chlordecone reductase; 3-alpha hydroxysteroid dehydrogenase, type I; dihydrodiol dehydrogenase 4; DD4; DD-4; Dihydrodiol dehydrogenase 4; dihydrodiol dehydrogenase isozyme DD4; HAKRA; HGNC:387; MGC22581; type I 3-alpha-hydroxysteroid dehydrogenase | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
1109 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P17516 | |
AKR1C4 | |
Synthetic peptide sequence (DPKYQRVELNDGH) corresponding to the N-terminus amino acids of AKR1C4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |