missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Albumin Monoclonal Antibody (AL-01), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119174
2203.44 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
This antibody will not cross-react with other mammalian species.
Human Serum Albumin Monoclonal antibody specifically detects Human Serum Albumin in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot.Specifications
Human Serum Albumin | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P02768 | |
ALB | |
Fraction of proteins containing albumin after ammonnium sulphate precipitation and DEAE-chromatography of human serum. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot | |
AL-01 | |
Unconjugated | |
ALB | |
Alb; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
Mouse | |
Protein A | |
RUO | |
213 | |
4° C, do not freeze | |
Liquid |