missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Synuclein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517820
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
alpha Synuclein Polyclonal antibody specifically detects alpha Synuclein in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
alpha Synuclein | |
Polyclonal | |
Unconjugated | |
SNCA | |
Alpha synuclein; alphaSYN; alpha-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Snca; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20617, 29219, 6622 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
60 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
O55042, P37377, P37840 | |
SNCA | |
Synthetic peptide corresponding to the carboxy-terminal sequence of human a-synuclein | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |