missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMIGO1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538045
Additional Details : Weight : 0.00900kg
Description
A recommended positive control is HeLa cell lysate. AMIGO1 antibody is not expected to cross-react with AMIGO2 or AMIGO3.
AMIGO1 Polyclonal antibody specifically detects AMIGO1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
AMIGO1 | |
Polyclonal | |
Unconjugated | |
Amigo1 | |
adhesion molecule with Ig like domain 1; adhesion molecule with Ig-like domain 1; ali2; alivin 2; alivin-2; AMIGO; AMIGO 1; AMIGO1; AMIGO-1; amigo1 {ECO:0000250; amphoretin-induced gene and ORF; amphoterin induced gene and ORF (Amigo); amphoterin-induced gene and ORF; amphoterin-induced protein 1; KIAA1163; MGC25558; OTTHUMP00000013379; RP23 89M15.6; transmembrane protein AMIGO; UniProtKB:Q80ZD8} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
229715, 295365, 57463 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q80ZD7, Q80ZD8, Q86WK6 | |
Amigo1 | |
19 amino acid peptide near the carboxy terminus of human AMIGO1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |