Learn More
BHMT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521470
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HepG2, mouse liver. Predicted reactivity: Mouse (95%), Rat (93%), Zebrafish (82%), Xenopus laevis (87%), Pig (95%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BHMT Polyclonal antibody specifically detects BHMT in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
BHMT | |
Polyclonal | |
Unconjugated | |
BHMT | |
betaine-homocysteine methyltransferase; betaine--homocysteine S-methyltransferase; betaine-homocysteine S-methyltransferase; betaine--homocysteine S-methyltransferase 1; BHMT; BHMT1; epididymis secretory sperm binding protein Li 61p; HEL-S-61p | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12116, 635 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O35490, Q93088 | |
BHMT | |
Recombinant fragment corresponding to a region within amino acids 1 and 243 of BHMT (Uniprot ID#Q93088) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |