missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Livin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA19065
Additional Details : Weight : 0.01000kg
Description
Livin Polyclonal antibody specifically detects Livin in Human samples. It is validated for Western Blot.Specifications
Livin | |
Polyclonal | |
Unconjugated | |
BIRC7 | |
baculoviral IAP repeat containing 7; baculoviral IAP repeat-containing 7; baculoviral IAP repeat-containing 7 (livin); baculoviral IAP repeat-containing protein 7; Baculoviral IAP repeat-containing protein 7 30 kDa subunit; Baculoviral IAP repeat-containing protein 7 30kDa subunit; Birc7; E130019N06; KIAP; Kidney inhibitor of apoptosis protein; LIVIN; livin inhibitor of apoptosis; melanoma inhibitor of apoptosis protein; MLIAP; ML-IAP; RGD1562883; RING finger protein 50; RING-type E3 ubiquitin transferase BIRC7; RNF50; RP11-261N11.7; Truncated livin; UNQ5800/PRO19607/PRO21344 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
79444 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96CA5 | |
BIRC7 | |
Synthetic peptide corresponding to the C terminus amino acids RAPVRSRVRT | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |