Learn More
Bmf Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534746
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: K562. Predicted reactivity: Mouse (87%), Rat (87%), Bovine (91%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Bmf Polyclonal antibody specifically detects Bmf in Human samples. It is validated for Western Blot.Specifications
Bmf | |
Polyclonal | |
Unconjugated | |
BMF | |
AW260063; AW411872; Bcl2 modifying factor; Bcl-2 modifying factor; bcl-2-modifying factor; Bmf; FLJ00065 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90427 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96LC9 | |
BMF | |
Recombinant fragment corresponding to a region within amino acids 1 and 184 of BMF (Uniprot ID#Q96LC9) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |