missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP-4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519683
Additional Details : Weight : 0.01000kg
Description
Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with cow and pig based on sequence homology.
BMP-4 Polyclonal antibody specifically detects BMP-4 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
BMP-4 | |
Polyclonal | |
Unconjugated | |
BMP4 | |
BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12159, 25296, 652 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.9 mg/mL | |
PBS with 0.02% sodium azide; pH 7.4 | |
P12644, P21275, Q06826 | |
BMP4 | |
Synthetic peptide conjugated to KLH derived from within residues 50 - 150 of Human BMP4. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |