Learn More
Bordetella pertussis LOS-A Monoclonal Antibody (D26E), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17005
Additional Details : Weight : 0.01000kg
Description
MA1-7005 detects Bordetella pertussis. This antibody is specific for LOS-A of Bordetella pertussis. MA1-7005 has been successfully used in ELISA, immunofluorescence and Western blot procedures. The MA1-7005 immunogen is whole B. pertussis lysate.
Bordetella pertussis LOS-A Monoclonal antibody specifically detects Bordetella pertussis LOS-A in Bacteria samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Bordetella pertussis LOS-A | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG3 |
ELISA, Immunocytochemistry, Western Blot | |
D26E | |
Unconjugated | |
B. pertussis; LOS-A | |
Whole Bordetella pertussis lysate. | |
1 mL | |
Primary | |
-20°C | |
Liquid |