missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA514787
Additional Details : Weight : 0.01000kg
Description
BRK Polyclonal antibody specifically detects BRK in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
BRK | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q13882 | |
PTK6 | |
KLH conjugated synthetic peptide between 11-41 amino acids from the N-terminal region of human BRK (PTK6) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PTK6 | |
ALT-PTK6; breast tumor kinase; BRK; deltam5; FLJ42088; protein tyrosine kinase 6; protein-tyrosine kinase 6; protein-tyrosine kinase BRK; Ptk6; PTK6 protein tyrosine kinase 6; Sik; SRC-related intestinal kinase; tks; Tksk; tyrosine kinase identified from skin; tyrosine-protein kinase BRK | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
5753 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |