missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
anti-C1QA Matched Antibody Pair, Abnova™
Marke: Abnova H00000712-AP21.1Set
Beinhaltet: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-C1QA (100μg); Detection antibody: mouse purified polyclonal anti-C1QA, (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Beschreibung
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human C1QA
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Spezifikation
| C1QA | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human C1QA. | |
| C1QA | |
| Affinity Purified | |
| RUO | |
| 712 | |
| Human |
| ELISA | |
| Unconjugated | |
| C1QA | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-C1QA (100 μg) 2. Detection antibody: mouse purified polyclonal anti-C1QA (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur