missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CK1 gamma-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538799
Additional Details : Weight : 0.00900kg
Description
Casein kinase I (also designated CKI) and casein kinase II (also designated CKII) compose a family of serine/ threonine protein kinases which are present in all eukaryotes examined to date. CKI family members, which include CKI alpha, gamma, epsilon and delta, have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. CKII is usually expressed as a tetrameric complex consisting of either an alpha2 beta2 or an alpha alpha′ beta2 structure. The alpha catalytic subunit is stimulated by the beta regulatory subunit, which undergoes autophosphorylation. CKII activity is high in the cytosol and nucleus of proliferating and differentiating cells. CKII is known to phosphorylate more than 100 different substrates including nuclear oncoproteins, transcription factors and enzymes involved in DNA metabolism.Specifications
CK1 gamma-1 | |
Polyclonal | |
Unconjugated | |
CSNK1G1 | |
Casein kinase I isoform gamma-1, CKI-gamma 1, CSNK1G1, KC1G1, CK1G1, CKI gamma1, CK-I gamma1, CKI- gamma1, CKI gamma-1, CKI gamma1, CK I gamma1, CKI gamma1, CKI gamma 1, CKI gamma 1, CKI-gamma-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
214897, 53944, 64086 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9HCP0, Q8BTH8, Q62761 | |
CSNK1G1 | |
A synthetic peptide derived from the N-terminal region of human CKI- gamma1 | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |