missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Eotaxin 2 Monoclonal Antibody (106521)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA523851
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.
Specifikationer
| Eotaxin 2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9JKC0 | |
| CCL24 | |
| E. coli-derived recombinant mouse CCL24/Eotaxin-2/MPIF-2 Ile24-Val119. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| ELISA, Neutralization, Western Blot | |
| 106521 | |
| Unconjugated | |
| CCL24 | |
| CC chemokine CCL24; C-C motif chemokine 24; C-C motif chemokine ligand 24; Ccl24; chemokine (C-C motif) ligand 24; Ckb-6; CK-beta-6; Eosinophil chemotactic protein 2; Eotaxin2; eotaxin-2; eotaxin-2-like protein; MPIF2; MPIF-2; Myeloid progenitor inhibitory factor 2; Scya24; small inducible cytokine A24; small inducible cytokine subfamily A (Cys-Cys), member 24; small-inducible cytokine A24 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 56221 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering