Läs mer
CD112 Mouse anti-Human, PE, Clone: R2.525, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551057
Beskrivning
Reacts with the poliovirus receptor 2 (PRR2) protein. PRR2 is a cell surface molecule homologous to the poliovirus receptor (PVR/CD155). It is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The PRR2 gene encodes for two mRNA isoforms, a short form of 3.0 kb (hPRR2 α ) and a long form of 4.4 kb (hPRR2 δ ). Both are widely expressed in human tissues, including hematopoietic cells. PRR2 is also expressed on endothelial cells, at the intercellular junctions of adjacent cells. It has been suggested that PRR2 may play a role in the regulation of hematopoietic/endothelial cell interaction.
Specifikationer
| CD112 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| R2.525 | |
| PRR2 | |
| Mouse | |
| 100 Tests | |
| Cell Biology | |
| Human | |
| IgG1 κ |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only.