missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11b Monoclonal Antibody (ICRF44), Alexa Fluor™ 488, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518124
Additional Details : Weight : 0.01000kg
Description
CD11b Monoclonal antibody specifically detects CD11b in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
CD11b | |
Monoclonal | |
Alexa Fluor 488 | |
ITGAM | |
antigen CD11b (p170); antigen CD11b (p170); macrophage antigen alpha polypeptide; CD11 antigen-like family member B; CD11b; CD11B (p170); CD11b/CD18; cell surface glycoprotein MAC-1 alpha subunit; cell surface glycoprotein MAC-1 subunit alpha; complement component 3 receptor 3 subunit; complement component receptor 3 alpha-a; complement receptor type 3; CR3; CR-3 alpha chain; CR3A; F730045J24Rik; integrin alpha M; integrin alpha-M; integrin subunit alpha M; integrin, alpha M; integrin, alpha M (complement component 3 receptor 3 subunit); ITGAM; Leukocyte adhesion receptor MO1; leukocyte integrin alpha-M chain; LOC100351865; Ly-40; MAC1; Mac-1; Mac-1 alpha; Mac-1 alpha (Mac1A); MAC-1 alpha subunit; MAC1A; Mac-1a; macrophage antigen alpha; macrophage antigen alpha polypeptide; MGC117044; MO1A; Neutrophil adherence receptor; neutrophil adherence receptor alpha-M subunit; SLEB6; unnamed protein product | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3684 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
ICRF44 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P11215 | |
ITGAM | |
Rheumatoid synovial cells and human monocytes | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |