Läs mer
PE-Cy™5 Mouse Anti-Human CD11b, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 555389
Beskrivning
The ICRF44 monoclonal antibody specifically binds to CD11b, the 165-kDa adhesion glycoprotein that associates with the 95-kDa integrin β2 (CD18) to form the CD11b/CD18 complex, also known as Mac-1 or CR3. CD11b is expressed on activated lymphocytes, monocytes, granulocytes, and a subset of NK cells. CD11b functions in cell-cell and cell-substrate interactions and is a receptor for iC3b, CD54 (ICAM-1), CD102 (ICAM-2) and CD50 (ICAM-3). This antibody significantly inhibits polymorphonuclear leukocyte aggregation in response to fMLP.
Specifikationer
| CD11b | |
| Monoclonal | |
| PE-Cyanine5 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human monocytes | |
| 100 Tests | |
| Immunology | |
| Human | |
| PE-Cy5 Mouse Anti-Human CD11b |
| Flow Cytometry | |
| ICRF44 | |
| Mac-1α, integrin αM subunit, CR3 α chain | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
| IgG1 κ |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only.