missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Monoclonal Antibody (BU15), APC, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA110084
Informations supplémentaires : Weight : 0.01000kg
Description
CD11c Monoclonal antibody specifically detects CD11c in Human, Non-human Primate samples. It is validated for Flow Cytometry.Spécification
CD11c | |
Monoclonal | |
APC | |
Itgax | |
AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3687 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
BU15 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P20702 | |
Itgax | |
Dendritic cells of synovial fluid | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |