Learn More
CD11c Monoclonal Antibody (BU15), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA182105
2683.46 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD11c antibody, clone Bu15 recognizes the integrin alphax subunit (ITGAX), a ~150 kDa glycoprotein also known as CD11 antigen-like family member C or Leu M5, CD11c is expressed by macrophages, monocytes, NK cells and most dendritic cells.
CD11c Monoclonal antibody specifically detects CD11c in Cynomolgus Monkey, Human samples. It is validated for Flow Cytometry.Specifications
CD11c | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
P20702 | |
Itgax | |
Proprietary Immunogen. | |
100 μg | |
Primary | |
Cynomolgus Monkey, Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
BU15 | |
FITC | |
Itgax | |
AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6 | |
Mouse | |
purified | |
RUO | |
102147272, 3687 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |