Läs mer
CD138 Rat anti-Mouse, Brilliant Violet 605, Clone: 281-2, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 563147
Beskrivning
The 281-2 monoclonal antibody specifically binds to the core protein of CD138 (Syndecan-1), a cell-surface, integral membrane heparan sulfate- and chondroitin sulfate-containing proteoglycan that binds to interstitial extracellular matrix molecules. Syndecan-1 is predominantly expressed on epithelial cells, where its expression correlates with normal epithelial organization. It is also expressed on B lymphocytes at specific stages during their differentiation: precursor B cells in the bone marrow and antibody-secreting cells, including plasma cells, but not mature peripheral B cells. It is thus implicated in mediating B cell-matrix interactions. CD138 expression is also regulated during embryonic development, and the molecule shows a tissue- specific structural polymorphism resulting from different post-translational modifications. The 281-2 antibody may be used to detect the differently glycosylated forms, because it reacts with the core protein. Furthermore, the mAb detects the Syndecan-1 ectodomain which is cleaved from cell surfaces by a metalloproteinase.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon™ BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon™ BV605 is a tandem fluorochrome of BD Horizon™ BV421 and Cy™3.5. Due to the excitation of Cy3.5 by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon™ PE-CF594 detectors off the green or yellow-green lasers. BD Horizon™ BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.
Specifikationer
| CD138 | |
| Monoclonal | |
| 0.2mg/mL | |
| SYND1; Syndecan-1; syn-1; Sdc1; Sstn; synstatin | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 281-2 | |
| Brilliant Violet 605 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| NAMRU mouse mammary gland epithelial cell line NMuMG | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.