Learn More
CD14 Monoclonal Antibody (5A3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514773
Additional Details : Weight : 0.01000kg
Description
MA5-14773 targets CD14 in indirect ELISA, FACS and IHC applications and shows reactivity with Human samples. The MA5-14773 immunogen is purified recombinant fragment of human CD14 expressed in E. Coli.
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry.Specifications
CD14 | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.03% sodium azide | |
P08571 | |
Cd14 | |
Purified recombinant fragment of human CD14 expressed in E. Coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry | |
5A3 | |
Unconjugated | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Mouse | |
Protein G | |
RUO | |
929 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |