Learn More
CD147 Monoclonal Antibody (MEM-M6/1), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA182332
Additional Details : Weight : 1.24740kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD147 antibody, clone MEM-M6/1 recognizes the human CD147 cell surface antigen, also known as Basigin, EMMPRIN and collagenase stimulatory factor.
CD147 Monoclonal antibody specifically detects CD147 in Human samples. It is validated for Flow Cytometry.Specifications
CD147 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4 | |
P35613 | |
BSG | |
CD147Rg, consisting of the cDNA coding for the hinge region, CH2 and CH3 domains of human IgG1. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
MEM-M6/1 | |
PE | |
BSG | |
5A11; 5A11 antigen; 5A11/Basigin; 5A11/Basigin-2; 5F7; AI115436; AI325119; Basic immunoglobulin superfamily; basigin; basigin (Ok blood group); Basigin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); basignin; Basignin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); blood-brain barrier HT7 antigen; BSG; CD147; CD147 antigen; CE9; Collagenase stimulatory factor; EMMPRIN; Extracellular matrix metalloproteinase inducer; glycoprotein CE9; gp 42; gp42/basigin/OX-47/HT7; HT7; HT-7; HT7 antigen; immunoglobulin gene superfamily member; leukocyte activation antigen M6; Membrane glycoprotein gp42; neurothelin; OK; OK blood group antigen; OX-47 antigen; Ox47R; RPE7; TCSF; tumor cell-derived collagenase stimulatory factor; Unknown (protein for MGC:128392); unnamed protein product; UNQ6505/PRO21383 | |
Mouse | |
Protein A | |
RUO | |
682 | |
4° C, do not freeze | |
Lyophilized |