missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA1 Monoclonal Antibody (MEM-158), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110106
Additional Details : Weight : 0.01000kg
Description
This antibody will not cross-react with porcine, bovine or sheep.
SSEA1 Monoclonal antibody specifically detects SSEA1 in Human samples. It is validated for Flow Cytometry.Specifications
SSEA1 | |
Monoclonal | |
APC | |
FUT4 | |
stage-specific embryonic antigen 1; CD15; SSEA-1; SSEA1; ELFT; FCT3A; FUC-TIV; FUTIV; LeX | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
2526 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-158 | |
TBS with 0.2% BSA and 15mM sodium azide; pH 8 | |
P22083 | |
FUT4 | |
Human granulocytes | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgM |