missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTLA-4 Monoclonal Antibody (WKH 203), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA170039
Additional Details : Weight : 0.01000kg
Description
MA1-70039 detects CD152 in rat samples. MA1-70039 has been successfully used in flow cytometry procedures. The MA1-70039 immunogen is purified rCTLA-4hlg fusion protein.
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Rat samples. It is validated for Flow Cytometry.Specifications
CTLA-4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
0 | |
CTLA4 | |
Purified rCTLA-4hlg fusion protein. | |
250 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
WKH 203 | |
Unconjugated | |
CTLA4 | |
ALPS5; CD; CD152; CD152 antigen; CD152 isoform; CD152 protein; CD152 protein precursor; celiac disease 3; CELIAC3; Ctla4; CTLA-4; cytotoxic T lymphocyte associated antigen 4 short spliced form; cytotoxic T-lymphocyte associated protein 4; cytotoxic T-lymphocyte protein 4; cytotoxic T-lymphocyte protein 4 isoform CTLA4-TM; cytotoxic T-lymphocyte-associated antigen 4; cytotoxic T-lymphocyte-associated protein 4; cytotoxic T-lymphocyte-associated serine esterase-4; EGK_04712; GRD4; GSE; ICOS; IDDM12; insulin-dependent diabetes mellitus 12; ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4; Ly-56; sCTLA4; soluble form; transmembrane form | |
Mouse | |
Protein G | |
RUO | |
63835 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |