Learn More
CD154 (CD40 Ligand) Monoclonal Antibody (MR-1), Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen MA110108
Additional Details : Weight : 0.01000kg
Description
Low endotoxin azide free phosphate buffered saline (PBS) solution, 0.2 μm filter sterilized. Endotoxin level is less than 0.01 EU/μg of the protein, as determined by the LAL test. Functional Application: Blocking.
CD154 (CD40 Ligand) Monoclonal antibody specifically detects CD154 (CD40 Ligand) in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.Specifications
CD154 (CD40 Ligand) | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P27548 | |
CD40LG | |
Activated mouse Th1 clone D1.6 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation | |
MR-1 | |
Unconjugated | |
CD40LG | |
CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5 | |
Armenian Hamster | |
Protein A | |
RUO | |
21947 | |
4° C, do not freeze | |
Liquid |