Learn More
BST-1 Monoclonal Antibody (KT157), FITC, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517948
1483.40 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
MA5-17948 targets CD157/BST1 in FACS applications and shows reactivity with Mouse samples. The MA5-17948 immunogen is t cell hybridoma J774.
BST-1 Monoclonal antibody specifically detects BST-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
BST-1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.02% sodium azide | |
Q64277 | |
Bst1 | |
T cell hybridoma J774 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2c |
Flow Cytometry | |
KT157 | |
FITC | |
Bst1 | |
114/A10; A530073F09; ADP-ribosyl cyclase 2; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2; antigen BP3; Bone marrow stromal antigen 1; bone marrow stromal cell antigen 1; Bp3; Bp-3; BP-3 alloantigen; Bst1; BST-1; Bsta1; cADPr hydrolase 2; CD157; CD157 antigen; Cyclic ADP-ribose hydrolase 2; leukocyte antigen 65; Ly65; Ly-65; NAD(+) nucleosidase; NAD+ nucleosidase | |
Rat | |
Protein G | |
RUO | |
12182 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |