missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD16 Monoclonal Antibody (3G8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110112
Additional Details : Weight : 0.01000kg
Description
Immunohistochemistry (Frozen): acetone fixation.
CD16 Monoclonal antibody specifically detects CD16 in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
CD16 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
O75015, P08637 | |
FCGR3A, FCGR3B | |
Human neutrophils | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
3G8 | |
Unconjugated | |
FCGR3A | |
4833442P21Rik; CD16; CD16-2; CD16A; CD16a antigen; CD16b; CD32; CD32 receptor 2; CDw32; cytolytic trigger molecule G7; Fc fragment of IgG intermediate affinity IV receptor; Fc fragment of IgG low affinity IIIa receptor; Fc fragment of IgG receptor IIIa; Fc fragment of IgG receptor IIIb; Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor; Fc fragment of IgG, low affinity IIIa, receptor (CD16a); Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIb, receptor (CD16b); Fc gamma receptor III; Fc gamma receptor IIIa; Fc gamma receptor III-A; Fc gamma receptor IIIb; Fc gamma RIIIa; Fc gammaRIV; Fc receptor, IgG, low affinity III; Fc receptor, IgG, low affinity IV; Fc receptor-like 3; Fcg receptor III; FCG2; FCG3; Fc-gamma receptor III-2 (CD 16); Fc-gamma receptor IIIb (CD 16); | |
Mouse | |
Protein A | |
RUO | |
2214, 2215 | |
4° C, do not freeze | |
Liquid |