missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRB1 Monoclonal Antibody (10/78), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516438
Additional Details : Weight : 0.01000kg
Description
KLRB1 Monoclonal antibody specifically detects KLRB1 in Rat samples. It is validated for Flow Cytometry.Specifications
KLRB1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
A4KWA1, P27471, Q0ZUP0 | |
Klrb1, Klrb1a, Klrb1b | |
Purified splenic NK cells from the LEW rat strain. | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
10/78 | |
FITC | |
Klrb1a | |
4930431A04Rik; Antigen 3.2.3; CD161; CD161 antigen-like family member A; CD161 antigen-like family member B; CD161a; CD161b; CLEC5B; C-type lectin domain family 5 member B; EMBL:AAQ11375.1}; Gm4696; HNKR-P1a; Immunoreceptor NKR-P1E; immunoreceptor NKR-P1G; inhibitory receptor NKR-P1B; killer cell lectin like receptor B1; killer cell lectin-like receptor subfamily A member 1 complex; killer cell lectin-like receptor subfamily B member 1; killer cell lectin-like receptor subfamily B member 1 complex; killer cell lectin-like receptor subfamily B member 1A; killer cell lectin-like receptor subfamily B member 1B; killer cell lectin-like receptor subfamily B member 1B allele A; Killer cell lectin-like receptor subfamily B member 1B allele B; killer cell lectin-like receptor subfamily B member 1B allele C; killer cell lectin-like receptor subfamily B member 1D; killer cell lectin-like receptor subfamily B member 1F; killer cell lectin-like receptor subfamily B member 1G; killer cell lectin-li | |
Mouse | |
Protein A | |
RUO | |
25192, 362443, 689817 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |