Learn More
CD166 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547083
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) ALCAM is observed, and less than 10% cross-reactivity with rhBCAM, recombinant mouse (rm) OCAM, and rmMAdCAM-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CD166 Polyclonal antibody specifically detects CD166 in Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.Specifications
CD166 | |
Polyclonal | |
Unconjugated | |
ALCAM | |
activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product | |
Goat | |
Antigen affinity chromatography | |
RUO | |
11658, 214, 478550, 79559 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O35112, O46634, Q13740, Q61490 | |
ALCAM | |
Mouse myeloma cell line NS0-derived recombinant mouse ALCAM/CD166 Trp28-Lys527 | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG |