Learn More
SIRP alpha Monoclonal Antibody (15-414), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA174228
Additional Details : Weight : 0.01000kg
Description
MA1-74228 is specific for human CD172a alpha. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD172a antibody, clone 15-414 recognizes human CD172a, also known as signal-regulatory protein alpha, a receptor-type transmembrane glycoprotein expressed on cells of myeloid origin, including granulocytes, dendritic cells (DCs), macrophages, mast cells and haematopoietic stem cells.
SIRP alpha Monoclonal antibody specifically detects SIRP alpha in Human samples. It is validated for Flow Cytometry.Specifications
SIRP alpha | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4 | |
P78324 | |
SIRPA | |
Monocyte-derived dendritic cells. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry | |
15-414 | |
PE | |
SIRPA | |
AI835480; Bit; brain Ig-like molecule with tyrosine-based activation motifs; Brain immunoglobulin like protein with tyrosine - based activation motifs; brain immunological-like with tyrosine-based motifs; brain-immunoglobulin-like molecule with tyrosine-based activation motifs; CD172; CD172 alpha; CD172 antigen-like family member A; CD172A; Inhibitory receptor SHPS-1; macrophage fusion receptor; Macrophage membrane protein MFP150; MFR; mSIRP-alpha1; Myd1; MYD-1; myD-1 antigen; p84; Protein tyrosine phosphatase non-receptor type substrate 1 (SHP substrate 1); protein tyrosine phosphatase, non-receptor type substrate 1; Protein tyrosine phosphatase, non-receptor type substrate 1 (SHP substrate 1); Ptpns1; SHP substrate 1; SHP-1; SHPS1; SHPS-1; signal regulatory protein alpha; signal-regulatory protein alpha; signal-regulatory protein alpha-1; Signal-regulatory protein alpha-2; Signal-regulatory protein alpha-3; SIRP; Sirpa; SIRPalpha; Sirp-alpha-1; SIRPalpha2; Sirp-alpha-2; Sirp-alpha-3; | |
Mouse | |
Protein G | |
RUO | |
140885 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |