missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD177 Monoclonal Antibody (MEM-166), Alexa Fluor™ 647, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518129
Additional Details : Weight : 0.01000kg
Description
CD177 Monoclonal antibody specifically detects CD177 in Human, Non-human Primate samples. It is validated for Flow Cytometry, Western Blot.Specifications
CD177 | |
Monoclonal | |
Alexa Fluor 647 | |
CD177 | |
1190003K14Rik; CD177; CD177 antigen; CD177 molecule; cell surface receptor; HNA2A; HNA-2a; Human neutrophil alloantigen 2a; NB1; NB1 glycoprotein; NB1 GP; Pdp3; Polycythemia rubra vera protein 1; PRV1; PRV-1; RGD1562941; UNQ595/PRO1181 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
57126 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry, Western Blot | |
MEM-166 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q8N6Q3 | |
CD177 | |
Human granulocytes | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |