Learn More
CD179a Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542149
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: TIRLTCTLRN DHDIGVYSVY WYQQRPGHPP RFLLRYFSQS DKSQGPQVPP Sequence homology: Cow: 92%; Dog: 86%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 79%
CD179a Polyclonal antibody specifically detects CD179a in Human samples. It is validated for Western Blot.Specifications
CD179a | |
Polyclonal | |
Unconjugated | |
VPREB1 | |
CD179 antigen-like family member A; CD179a; IGI; IGLL1; IGVPB; Immunoglobulin iota chain; Immunoglobulin lambda Vpreb1 chain; immunoglobulin lambda-1 light chain; immunoglobulin lambda-like polypeptide 5; LOC100735142; pre-B lymphocyte 1; pre-B lymphocyte gene 1; Protein VPreB1; surrogate light chain; v(pre)B protein; VPREB; VpreB protein; Vpreb1; Vpreb-1; V-set pre-B cell surrogate light chain 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7441 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P12018 | |
VPREB1 | |
Synthetic peptide directed towards the middle region of human VPREB1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |