missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD18 Monoclonal Antibody (MEM-148), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119469
2683.46 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
This antibody will not cross-react with porcine.
CD18 Monoclonal antibody specifically detects CD18 in Human samples. It is validated for Flow Cytometry.Specifications
CD18 | |
Monoclonal | |
APC | |
ITGB2 | |
2E6; AI528527; antigen CD18; B-2 integrin; CD18; CD18 leukocyte adhesion molecule; CD18 subunit; cell surface adhesion glycoprotein (LFA-1/CR3/P150,959 beta subunit precursor); cell surface adhesion glycoproteins LFA-1/CR3/p150,95 subunit beta; complement component 3 receptor 3 and 4 subunit; complement receptor C3 beta-subunit; complement receptor C3 subunit beta; integrin beta 2; integrin beta chain, beta 2; integrin beta-2; integrin subunit beta 2; integrin, beta 2; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; macrophage antigen 1 (mac-1) beta subunit); integrin, beta 2 (antigen CD18 subunit (p95), lymphocyte function-associated antigen 1, integrin B2); integrin, beta 2 (complement component 3 receptor 3 and 4 subunit); ITGB2; LAD; LCAMB; Leu-CAM receptor; leukocyte cell adhesion molecule CD18; leukocyte surface protein; leukocyte-associated antigens CD18/11A, CD18/11B, | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3689 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-148 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P05107 | |
ITGB2 | |
Peripheral blood mononuclear cells. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |