Learn More
CD180 Monoclonal Antibody (RP/14), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA140034
Additional Details : Weight : 0.01000kg
Description
MA1-40034 detects CD180 from mouse samples. MA1-40034 has been successfully used in flow cytometry and funtional applications. The MA1-40034 immunogen is mouse RP105/CD180. Endotoxin level < 24 EU/mg.
CD180 Monoclonal antibody specifically detects CD180 in Mouse samples. It is validated for Flow Cytometry, Functional Assay.Specifications
CD180 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and no preservative | |
Q62192 | |
Cd180 | |
Murine RP105/CD180. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry, Functional Assay | |
RP/14 | |
Unconjugated | |
Cd180 | |
CD180; CD180 antigen; CD180 molecule; F630107B15; LY64; Ly78; Ly-78; lymphocyte antigen 64; Lymphocyte antigen 78; lymphocyte antigen-64, radioprotective, 105kDa; MGC126233; MGC126234; radioprotective 105 kDa protein; RP105 | |
Rat | |
Protein G | |
RUO | |
17079 | |
4° C | |
Liquid |