Learn More
CD1a Monoclonal Antibody (SP157), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA516333
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 1mM EDTA buffer, pH 8.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is thymus or skin tissue.
CD1a Monoclonal antibody specifically detects CD1a in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
CD1a | |
Monoclonal | |
Unconjugated | |
CD1A | |
CD1; CD1 antigen; CD1.1; CD1A; CD1A antigen, a polypeptide; CD1a molecule; cd1a1; CD1A1 antigen; cluster of differentiation 1 A; cortical thymocyte antigen CD1A; differentiation antigen CD1-alpha-3; epidermal dendritic cell marker CD1a; FCB6; HTA1; hTa1 thymocyte antigen; R4; T6; T-cell surface antigen T6/Leu-6; T-cell surface glycoprotein CD1a | |
Rabbit | |
Protein A/G | |
RUO | |
909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
SP157 | |
PBS with 1% BSA and <0.1% sodium azide; pH 7.6 | |
P06126 | |
CD1A | |
Synthetic peptide derived from the internal region of human CD1a protein. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |