Learn More
CD1d Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521625
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa, Molt-4, Raji. Predicted reactivity: Rhesus Monkey (91%), Chimpanzee (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD1d Polyclonal antibody specifically detects CD1d in Human samples. It is validated for Western Blot.Specifications
CD1d | |
Polyclonal | |
Unconjugated | |
CD1D | |
AI747460; Antigen-presenting glycoprotein CD1d; antigen-presenting glycoprotein CD1d1; Cd1; CD1.1; CD1A; Cd1d; CD1D antigen; CD1D antigen, d polypeptide; CD1d molecule; CD1d.1; Cd1d1; CD1d1 antigen; CD1d1 molecule; differentiation antigen CD1-alpha-3; HMC class I antigen-like glycoprotein CD1D; Ly-38; R3; R3G1; T-cell surface glycoprotein CD1d; T-cell surface glycoprotein CD1d1; thymocyte antigen CD1D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
912 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.91 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P15813 | |
CD1D | |
Recombinant fragment corresponding to a region within amino acids 56 and 317 of CD1D (Uniprot ID#P15813) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |